Assay Details
Target Gene Details
Entrez Gene ID: | 844 |
Gene Name: | calsequestrin 1 |
Gene Aliases: |
CASQ, PDIB1, VMCQA |
Location: |
Chr.1:160190495-160201886 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASQ1 | NM_001231.4 | NP_001222.3 | ||
| AK300713.1 | ||||
| AK313250.1 | ||||
| BC022289.1 | AAH22289.1 | |||
| S73775.1 | AAB32063.1 |
More Information
Set Membership: |
|

Genomic Map