Assay Details
Target Gene Details
Entrez Gene ID: | 9033 |
Gene Name: | polycystin 2 like 1, transient receptor potential cation channel |
Gene Aliases: |
PCL, PKD2L, PKDL, TRPP3 |
Location: |
Chr.10:100288145-100346472 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKD2L1 | NM_001253837.1 | 7 | 1678 | NP_001240766.1 |
| NM_016112.2 | 7 | 1656 | NP_057196.2 | |
| XM_011540323.2 | 8 | 1219 | XP_011538625.1 | |
| XM_011540325.2 | 9 | 1329 | XP_011538627.1 | |
| XM_017016875.1 | 8 | 1167 | XP_016872364.1 | |
| XM_017016876.1 | 6 | 925 | XP_016872365.1 | |
| AF053316.1 | 7 | 1148 | AAD51859.1 | |
| AF073481.1 | 7 | 1656 | AAD41638.1 | |
| AF092170.1 | 5 | 935 | AAD16101.1 | |
| AF094827.1 | 6 | 988 | AAD08695.1 | |
| BC025665.1 | 7 | 1353 | AAH25665.1 | |
| DQ084244.1 | 7 | 1132 | AAZ31363.1 |
More Information
Set Membership: |
|

Genomic Map