Assay Details
Target Gene Details
Entrez Gene ID: | 8567 |
Gene Name: | MAP kinase activating death domain |
Gene Aliases: |
DENN, IG20, RAB3GEP |
Location: |
Chr.11:47269376-47330031 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
More Information
Set Membership: |
|

Genomic Map