Assay Details
Target Gene Details
Entrez Gene ID: | 155 |
Gene Name: | adrenoceptor beta 3 |
Gene Aliases: |
BETA3AR |
Location: |
Chr.8:37962995-37966666 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRB3 | NM_000025.2 | 1 | 426 | NP_000016.1 |
| AK293033.1 | 1 | 370 | ||
| AK309919.1 | 1 | 357 | ||
| AK313857.1 | 1 | 354 | ||
| AY487247.1 | 1 | 229 | AAR37414.1 | |
| BC075017.2 | 1 | 268 | AAH75017.1 | |
| FJ170286.1 | 1 | 229 | ||
| FJ170287.1 | 1 | 229 | ||
| X70811.1 | 1 | 354 | CAA50141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610947 | Chr.8:37935047 - 38054626 on Build GRCh38 | Loss |
|
| nsv610948 | Chr.8:37963968 - 37967299 on Build GRCh38 | Loss |
|
| nsv951127 | Chr.8:37964383 - 37967082 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map