Assay Details
Target Gene Details
Entrez Gene ID: | 79837 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase type 2 gamma |
Gene Aliases: |
PIP5K2C |
Location: |
Chr.12:57591159-57603428 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP4K2C | NM_001146258.1 | NP_001139730.1 | ||
| NM_001146259.1 | NP_001139731.1 | |||
| NM_001146260.1 | NP_001139732.1 | |||
| NM_024779.4 | NP_079055.3 | |||
| XM_005269152.2 | XP_005269209.1 | |||
| XM_011538747.1 | XP_011537049.1 | |||
| XM_017019973.1 | XP_016875462.1 | |||
| AK025708.1 | BAB15223.1 | |||
| AK075553.1 | ||||
| AK125526.1 | ||||
| AK225343.1 | ||||
| AK225685.1 | ||||
| AK297243.1 | ||||
| AK302254.1 | ||||
| AK315588.1 | ||||
| AX754970.1 | CAD99241.1 | |||
| BC028596.2 | AAH28596.1 | |||
| BX419431.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428283 | Chr.12:57435950 - 57608085 on Build GRCh38 | Gain |
|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv725 | Chr.12:57572766 - 57600025 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map