Assay Details
Target Gene Details
Entrez Gene ID: | 7043 |
Gene Name: | transforming growth factor beta 3 |
Gene Aliases: |
ARVD, ARVD1, LDS5, RNHF, TGF-beta3 |
Location: |
Chr.14:75958097-75982022 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB3 | NM_003239.3 | NP_003230.1 | ||
| AK122902.1 | 1 | 1055 | ||
| AK313564.1 | ||||
| BC018503.1 | AAH18503.1 | |||
| BT007287.1 | AAP35951.1 | |||
| EF560714.1 | ||||
| J03241.1 | AAA61161.1 | |||
| X14149.1 | CAA32362.1 |
More Information
Set Membership: |
|

Genomic Map