Assay Details
Target Gene Details
Entrez Gene ID: | 3429 |
Gene Name: | interferon alpha inducible protein 27 |
Gene Aliases: |
FAM14D, ISG12, ISG12A, P27 |
Location: |
Chr.14:94110733-94116699 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFI27 | NM_001130080.2 | NP_001123552.1 | ||
| NM_001288952.1 | NP_001275881.1 | |||
| NM_001288954.1 | NP_001275883.1 | |||
| NM_001288956.1 | NP_001275885.1 | |||
| NM_001288957.1 | NP_001275886.1 | |||
| NM_001288958.1 | NP_001275887.1 | |||
| NM_001288959.1 | NP_001275888.1 | |||
| NM_001288960.1 | NP_001275889.1 | |||
| NM_001288995.1 | NP_001275924.1 | |||
| NM_005532.4 | NP_005523.3 | |||
| AI799552.1 | ||||
| AK095039.1 | ||||
| AK289535.1 | ||||
| AK312089.1 | ||||
| BC015492.1 | AAH15492.1 | |||
| BF971643.1 | ||||
| BG483267.1 | ||||
| BM756566.1 | ||||
| BM841506.1 | ||||
| BM841596.1 | ||||
| BP317624.1 | ||||
| BT006781.1 | AAP35427.1 | |||
| BU601451.1 | ||||
| BU688913.1 | ||||
| BU928223.1 | ||||
| BX647330.1 | ||||
| CB993374.1 | ||||
| X67325.1 | CAA47739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892577 | Chr.14:94096847 - 94118727 on Build GRCh38 | Gain |
|
| esv34067 | Chr.14:94024355 - 94160851 on Build GRCh38 | Loss |
|
| esv3635373 | Chr.14:94040151 - 94173989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map