Assay Details
Target Gene Details
Entrez Gene ID: | 58190 |
Gene Name: | CTD small phosphatase 1 |
Gene Aliases: |
NIF3, NLI-IF, NLIIF, SCP1 |
Location: |
Chr.2:218398338-218405941 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTDSP1 | NM_001206878.1 | 7 | 1503 | NP_001193807.1 |
| NM_021198.2 | 7 | 1767 | NP_067021.1 | |
| NM_182642.2 | 7 | 1764 | NP_872580.1 | |
| XM_011511569.2 | 7 | 1914 | XP_011509871.1 | |
| XM_011511570.2 | 7 | 1910 | XP_011509872.1 | |
| AF217969.1 | 5 | 1171 | AAG17212.1 | |
| AF229162.1 | 7 | 1767 | AAG15402.1 | |
| AF258582.1 | 5 | 1227 | AAG23785.1 | |
| AK075090.1 | 2 | 896 | ||
| BC012977.2 | 7 | 1476 | AAH12977.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3166 | Chr.2:218371765 - 218405183 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map