Assay Details
Target Gene Details
Entrez Gene ID: | 5074 |
Gene Name: | pro-apoptotic WT1 regulator |
Gene Aliases: |
PAR4, Par-4 |
Location: |
Chr.12:79587791-79691097 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAWR | NM_002583.2 | 7 | 1592 | NP_002574.2 |
| XM_006719435.2 | 7 | 1568 | XP_006719498.1 | |
| XM_017019377.1 | 7 | 1472 | XP_016874866.1 | |
| BC007018.1 | 6 | 1438 | AAH07018.1 | |
| CA313080.1 | ||||
| U63809.1 | 7 | 1550 | AAC24947.1 |
More Information
Set Membership: |
|

Genomic Map