Assay Details
Target Gene Details
Entrez Gene ID: | 51281 |
Gene Name: | ankyrin repeat and MYND domain containing 1 |
Gene Aliases: |
ZMYND13 |
Location: |
Chr.2:240468632-240561110 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759119 | Chr.2:240425704 - 240649454 on Build GRCh38 | Gain |
|
| nsv997288 | Chr.2:239542278 - 240534009 on Build GRCh38 | Gain |
|
| nsv508899 | Chr.2:240333579 - 240490606 on Build GRCh38 | Insertion |
|
| nsv3239 | Chr.2:240476823 - 240510855 on Build GRCh38 | Insertion |
|
| nsv834586 | Chr.2:240475181 - 240622625 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map