Assay Details
Target Gene Details
Entrez Gene ID: | 7158 |
Gene Name: | tumor protein p53 binding protein 1 |
Gene Aliases: |
53BP1, TDRD30, TP53, p202, p53BP1 |
Location: |
Chr.15:43403064-43510509 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP1 | NM_001141979.1 | NP_001135451.1 | ||
| NM_001141980.1 | NP_001135452.1 | |||
| NM_005657.2 | NP_005648.1 | |||
| XM_011521985.1 | XP_011520287.1 | |||
| XM_011521986.2 | XP_011520288.1 | |||
| XM_017022533.1 | XP_016878022.1 | |||
| AB210025.1 | ||||
| AF078776.1 | AAC62018.1 | |||
| AK295764.1 | ||||
| BC112161.1 | AAI12162.1 | |||
| BX537418.1 | CAD97660.1 | |||
| U09477.1 | AAA21596.1 |
More Information
Set Membership: |
|

Genomic Map