Assay Details
Target Gene Details
Entrez Gene ID: | 2634 |
Gene Name: | guanylate binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.1:89106132-89126159 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP2 | NM_004120.4 | NP_004111.2 | ||
| AK057968.1 | ||||
| AK226018.1 | ||||
| AK314325.1 | ||||
| BC022272.1 | AAH22272.1 | |||
| BC073163.1 | AAH73163.1 | |||
| CA390345.1 | ||||
| CR457062.1 | CAG33343.1 | |||
| DB274428.1 | ||||
| M55543.1 | AAA67323.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586753 | Chr.1:89119990 - 89126777 on Build GRCh38 | Loss |
|
| nsv546797 | Chr.1:89081456 - 89123873 on Build GRCh38 | Loss |
|
| nsv546796 | Chr.1:89052437 - 89256179 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map