Assay Details
Target Gene Details
Entrez Gene ID: | 11010 |
Gene Name: | GLI pathogenesis related 1 |
Gene Aliases: |
CRISP7, GLIPR, RTVP1 |
Location: |
Chr.12:75480733-75501936 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLIPR1 | NM_006851.2 | NP_006842.2 | ||
| CA421777.1 |
Target Gene Details
Entrez Gene ID: | 11103 |
Gene Name: | KRR1, small subunit processome component homolog |
Gene Aliases: |
HRB2, RIP-1 |
Location: |
Chr.12:75497639-75511638 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRR1 | NM_007043.6 | NP_008974.5 | ||
| AK313687.1 | ||||
| BC005225.1 | ||||
| BC016778.1 | AAH16778.1 | |||
| BC026107.2 | AAH26107.1 | |||
| BC033887.2 | AAH33887.1 | |||
| EF010919.1 | ||||
| U55766.1 | AAB00557.1 |
More Information
Set Membership: |
|

Genomic Map