Assay Details
Target Gene Details
Entrez Gene ID: | 203 |
Gene Name: | adenylate kinase 1 |
Gene Aliases: |
HTL-S-58j |
Location: |
Chr.9:127866480-127886749 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK1 | NM_000476.2 | 7 | 1907 | NP_000467.1 |
| NM_001318121.1 | 7 | 1894 | NP_001305050.1 | |
| NM_001318122.1 | 6 | 1902 | NP_001305051.1 | |
| EU794628.1 | 7 | 1907 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28616 | Chr.9:127860289 - 127884215 on Build GRCh38 | Gain+Loss |
|
| esv3621795 | Chr.9:127863894 - 127881499 on Build GRCh38 | Gain |
|
| esv3576708 | Chr.9:127866309 - 127882824 on Build GRCh38 | Gain |
|
| nsv466575 | Chr.9:127770709 - 127866860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map