Assay Details
Target Gene Details
Entrez Gene ID: | 150 |
Gene Name: | adrenoceptor alpha 2A |
Gene Aliases: |
ADRA2, ADRA2R, ADRAR, ALPHA2AAR, ZNF32 |
Location: |
Chr.10:111077032-111080907 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRA2A | NM_000681.3 | 1 | 673 | NP_000672.3 |
| AF284095.1 | 1 | 542 | AAK26743.1 | |
| BC050414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825566 | Chr.10:111070256 - 111111575 on Build GRCh38 | Gain |
|
| nsv527400 | Chr.10:111073803 - 111083327 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map