Assay Details
Target Gene Details
Entrez Gene ID: | 4091 |
Gene Name: | SMAD family member 6 |
Gene Aliases: |
AOVD2, HsT17432, MADH6, MADH7 |
Location: |
Chr.15:66702110-66782000 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD6 | NM_005585.4 | 1 | 932 | NP_005576.3 |
| NR_027654.1 | 1 | 932 | ||
| AF035528.1 | 1 | 949 | AAB94137.1 | |
| AF037469.1 | AAC82331.1 | |||
| AF043640.1 | AAC00497.1 | |||
| AM909653.1 | ||||
| BC012986.2 | 1 | 62 | AAH12986.1 | |
| CB055216.1 | 1 | 155 |
More Information
Set Membership: |
|

Genomic Map