Assay Details
Target Gene Details
Entrez Gene ID: | 8431 |
Gene Name: | nuclear receptor subfamily 0 group B member 2 |
Gene Aliases: |
SHP, SHP1 |
Location: |
Chr.1:26911484-26914076 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR0B2 | NM_021969.2 | 2 | 1139 | NP_068804.1 |
| AA872285.1 | 1 | 139 | ||
| BC030207.1 | 2 | 1029 | AAH30207.1 |
Target Gene Details
Entrez Gene ID: | 10726 |
Gene Name: | nuclear distribution C, dynein complex regulator |
Gene Aliases: |
HNUDC, MNUDC, NPD011 |
Location: |
Chr.1:26911128-26946871 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDC | XM_011540529.1 | XP_011538831.1 | ||
| XM_017000094.1 | XP_016855583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000012 | Chr.1:26714969 - 27141528 on Build GRCh38 | Gain |
|
| nsv834524 | Chr.1:26830184 - 26994502 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map