Assay Details
Target Gene Details
Entrez Gene ID: | 5875 |
Gene Name: | Rab geranylgeranyltransferase alpha subunit |
Gene Aliases: |
PTAR3 |
Location: |
Chr.14:24265538-24271627 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RABGGTA | NM_004581.5 | NP_004572.3 | ||
| NM_182836.2 | NP_878256.1 | |||
| AK291347.1 | ||||
| AK292613.1 | ||||
| BC003093.1 | AAH03093.1 | |||
| CD370056.1 | ||||
| CR541797.1 | CAG46596.1 | |||
| Y08200.1 | CAA69382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952818 | Chr.14:24249495 - 24270894 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map