Assay Details
Target Gene Details
Entrez Gene ID: | 4682 |
Gene Name: | nucleotide binding protein 1 |
Gene Aliases: |
NBP, NBP1, NBP35 |
Location: |
Chr.16:10743164-10769351 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUBP1 | NM_001278506.1 | NP_001265435.1 | ||
| NM_001323594.1 | NP_001310523.1 | |||
| NM_001323595.1 | NP_001310524.1 | |||
| NM_001323596.1 | NP_001310525.1 | |||
| NM_001323597.1 | NP_001310526.1 | |||
| NM_002484.3 | NP_002475.2 | |||
| XM_017023252.1 | XP_016878741.1 | |||
| AK223204.1 | BAD96924.1 | |||
| AK307148.1 | ||||
| AK310089.1 | ||||
| BC100290.1 | AAI00291.1 | |||
| BC109322.1 | AAI09323.1 | |||
| BC109323.1 | AAI09324.1 | |||
| BP216107.1 | ||||
| BP225034.1 | ||||
| U01833.1 | AAA61932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833140 | Chr.16:10664013 - 10791497 on Build GRCh38 | Loss |
|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map