Assay Details
Target Gene Details
Entrez Gene ID: | 55628 |
Gene Name: | zinc finger protein 407 |
Gene Aliases: |
- |
Location: |
Chr.18:74597735-75065672 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF407 | NM_001146189.1 | 1 | 3288 | NP_001139661.1 |
| NM_001146190.1 | 1 | 3288 | NP_001139662.1 | |
| NM_017757.2 | 1 | 3288 | NP_060227.2 | |
| XM_005266726.4 | 2 | 3487 | XP_005266783.1 | |
| XM_006722500.3 | 2 | 3352 | XP_006722563.1 | |
| XM_011526068.2 | 2 | 6994 | XP_011524370.1 | |
| XM_011526069.2 | 2 | 3352 | XP_011524371.1 | |
| XM_011526070.1 | 2 | 3352 | XP_011524372.1 | |
| XM_017025838.1 | 2 | 3352 | XP_016881327.1 | |
| AB051490.1 | BAB21794.1 | |||
| AK000314.1 | 1 | 913 | BAA91077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2357 | Chr.18:74633579 - 74667562 on Build GRCh38 | Insertion |
|
| nsv1057441 | Chr.18:74344906 - 74669985 on Build GRCh38 | Gain |
|
| nsv577656 | Chr.18:74534683 - 75005167 on Build GRCh38 | Gain |
|
| esv3643124 | Chr.18:74623637 - 74695240 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map