Assay Details
Target Gene Details
Entrez Gene ID: | 7011 |
Gene Name: | telomerase associated protein 1 |
Gene Aliases: |
TLP1, TP1, TROVE1, VAULT2, p240 |
Location: |
Chr.14:20365667-20413540 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEP1 | NM_001319035.1 | 39 | 5749 | NP_001305964.1 |
| NM_007110.4 | 41 | 6073 | NP_009041.2 | |
| XM_005268027.4 | 41 | 6213 | XP_005268084.1 | |
| XM_011537110.2 | 33 | 4558 | XP_011535412.1 | |
| AB209669.1 | 29 | 4059 | BAD92906.1 | |
| AK303307.1 | 12 | 1665 | ||
| BC126107.1 | 41 | 6073 | ||
| U86136.2 | 41 | 6073 | AAC51107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952812 | Chr.14:20371342 - 20387141 on Build GRCh38 | Deletion |
|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
| nsv563846 | Chr.14:20376700 - 20386079 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map