Assay Details
Target Gene Details
Entrez Gene ID: | 26577 |
Gene Name: | procollagen C-endopeptidase enhancer 2 |
Gene Aliases: |
PCPE2 |
Location: |
Chr.3:142817860-142889203 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCOLCE2 | NM_013363.3 | NP_037495.1 | ||
| AF098269.1 | AAF04621.1 | |||
| AK075478.1 | ||||
| AK223633.1 | BAD97353.1 | |||
| AK315121.1 | ||||
| AY035400.1 | AAK63128.1 | |||
| AY358557.1 | AAQ88921.1 | |||
| CR536570.1 | CAG38807.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000631 | Chr.3:142797525 - 142853216 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map