Assay Details
Target Gene Details
Entrez Gene ID: | 55789 |
Gene Name: | DEP domain containing 1B |
Gene Aliases: |
BRCC3, XTP1 |
Location: |
Chr.5:60596912-60700166 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEPDC1B | NM_001145208.1 | 10 | 1488 | NP_001138680.1 |
| NM_018369.2 | 11 | 1674 | NP_060839.2 | |
| XM_011543509.2 | 11 | 1667 | XP_011541811.1 | |
| AF303178.1 | 11 | 1642 | AAN31126.1 | |
| AF488828.1 | AAO49477.1 | |||
| AF490257.1 | AAO85465.1 | |||
| AK001166.1 | 4 | 585 | BAA91529.1 | |
| AK290684.1 | 11 | 1669 | ||
| AK299242.1 | 11 | 1522 | ||
| AK300782.1 | 10 | 1456 | ||
| BC010904.1 | 5 | 742 | AAH10904.1 | |
| BC019075.1 | 11 | 1637 | AAH19075.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018614 | Chr.5:60412396 - 60614481 on Build GRCh38 | Loss |
|
| nsv598288 | Chr.5:60331728 - 60707633 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map