Assay Details
Target Gene Details
Entrez Gene ID: | 5420 |
Gene Name: | podocalyxin like |
Gene Aliases: |
Gp200, PC, PCLP, PCLP-1 |
Location: |
Chr.7:131500262-131556617 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PODXL | NM_001018111.2 | NP_001018121.1 | ||
| NM_005397.3 | NP_005388.2 | |||
| AK055816.1 | BAB71022.1 | |||
| AK223573.1 | BAD97293.1 | |||
| BC093730.1 | AAH93730.1 | |||
| BC112035.1 | AAI12036.1 | |||
| BC143318.1 | ||||
| BG006197.1 | ||||
| CN290641.1 | ||||
| U97519.1 | AAB61574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608424 | Chr.7:131506702 - 131562661 on Build GRCh38 | Loss |
|
| nsv831134 | Chr.7:131455230 - 131614283 on Build GRCh38 | Gain |
|
| dgv6617n100 | Chr.7:131331684 - 131555085 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map