Assay Details
Target Gene Details
Entrez Gene ID: | 10042 |
Gene Name: | HMG-box containing 4 |
Gene Aliases: |
HMG2L1, HMGBCG, THC211630 |
Location: |
Chr.22:35257274-35295807 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGXB4 | NM_001003681.2 | NP_001003681.1 | ||
| NR_027780.1 | ||||
| XM_006724100.3 | XP_006724163.1 | |||
| XM_006724101.3 | XP_006724164.1 | |||
| XM_006724102.1 | XP_006724165.1 | |||
| AJ010070.1 | CAA08992.1 | |||
| AK299833.1 | ||||
| AL079310.1 | CAB45240.1 | |||
| BC004194.2 | AAH04194.1 | |||
| BC017326.2 | AAH17326.1 | |||
| BC025303.2 | AAH25303.1 | |||
| BC052624.1 | AAH52624.1 | |||
| BC062413.1 | AAH62413.1 | |||
| BC111373.1 | AAI11374.1 | |||
| CB991207.1 | ||||
| CR456504.1 | CAG30390.1 | |||
| DW452042.1 |
More Information
Set Membership: |
|

Genomic Map