Assay Details
Target Gene Details
Entrez Gene ID: | 79760 |
Gene Name: | gem nuclear organelle associated protein 7 |
Gene Aliases: |
SIP3 |
Location: |
Chr.19:45075728-45091524 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GEMIN7 | NM_001007269.1 | NP_001007270.1 | ||
| NM_001007270.1 | NP_001007271.1 | |||
| NM_001319054.1 | NP_001305983.1 | |||
| NM_001319055.1 | NP_001305984.1 | |||
| NM_024707.2 | NP_078983.1 | |||
| XM_005259262.3 | XP_005259319.1 | |||
| XM_005259263.4 | XP_005259320.1 | |||
| XM_017027312.1 | XP_016882801.1 | |||
| AK024018.1 | BAB14780.1 | |||
| AW297699.1 | ||||
| AY114106.1 | AAM44083.1 | |||
| BC007793.2 | AAH07793.1 | |||
| BM009097.1 | ||||
| BU602037.1 | ||||
| CB148876.1 | ||||
| CR457321.1 | CAG33602.1 | |||
| W35331.1 |
Target Gene Details
Entrez Gene ID: | 105372419 |
Gene Name: | uncharacterized LOC105372419 |
Gene Aliases: |
- |
Location: |
Chr.19:45085329-45090391 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105372419 | NR_134887.1 | |||
| AL047654.1 | ||||
| BI087770.1 | ||||
| CN478580.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953587 | Chr.19:45039143 - 45166142 on Build GRCh38 | Deletion |
|
| esv3644497 | Chr.19:45085975 - 45092273 on Build GRCh38 | Loss |
|
| esv27359 | Chr.19:45085653 - 45092119 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map