Assay Details
Target Gene Details
Entrez Gene ID: | 441168 |
Gene Name: | family with sequence similarity 26 member F |
Gene Aliases: |
C6orf187, dJ93H18.5 |
Location: |
Chr.6:116461370-116463782 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM26F | NM_001010919.2 | NP_001010919.1 | ||
| NM_001276460.1 | NP_001263389.1 | |||
| XM_005266999.3 | 1 | 370 | XP_005267056.1 | |
| XM_011535845.2 | 1 | 384 | XP_011534147.1 | |
| AV734646.2 | ||||
| BC146842.1 | ||||
| BM704529.1 | ||||
| DN997293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830783 | Chr.6:116445706 - 116645069 on Build GRCh38 | Gain |
|
| nsv523972 | Chr.6:115958024 - 116809541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map