Assay Details
Target Gene Details
Entrez Gene ID: | 26266 |
Gene Name: | solute carrier family 13 member 4 |
Gene Aliases: |
NAS2, SUT-1, SUT1 |
Location: |
Chr.7:135681237-135728204 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC13A4 | NM_001318192.1 | 8 | 1527 | NP_001305121.1 |
| NM_012450.3 | 8 | 1524 | NP_036582.2 | |
| XM_011516024.2 | 8 | 1307 | XP_011514326.1 | |
| XM_017011962.1 | 6 | 895 | XP_016867451.1 | |
| XM_017011963.1 | 8 | 1307 | XP_016867452.1 | |
| AB208809.1 | 5 | 5802 | BAD92046.1 | |
| AF169301.1 | 8 | 1497 | AAF05907.1 | |
| AK313072.1 | 8 | 1117 | ||
| BC030689.2 | 8 | 1506 | AAH30689.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608460 | Chr.7:135434669 - 135798303 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map