Assay Details
Target Gene Details
Entrez Gene ID: | 8772 |
Gene Name: | Fas associated via death domain |
Gene Aliases: |
GIG3, MORT1 |
Location: |
Chr.11:70203163-70207402 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FADD | NM_003824.3 | 1 | 351 | NP_003815.1 |
| AK291005.1 | 1 | 212 | ||
| AK293320.1 | 1 | 208 | ||
| AL575732.1 | 1 | 351 | ||
| AY423721.1 | AAS00484.1 | |||
| BC000334.2 | 1 | 199 | AAH00334.1 | |
| BT006927.1 | AAP35573.1 | |||
| CR456738.1 | CAG33019.1 | |||
| U24231.1 | 1 | 183 | AAA86517.1 | |
| X84709.1 | 1 | 198 | CAA59197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555376 | Chr.11:70156780 - 70248052 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map