Assay Details
Target Gene Details
Entrez Gene ID: | 59341 |
Gene Name: | transient receptor potential cation channel subfamily V member 4 |
Gene Aliases: |
BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC |
Location: |
Chr.12:109783087-109833407 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPV4 | NM_001177428.1 | 9 | 1667 | NP_001170899.1 |
| NM_001177431.1 | 11 | 1675 | NP_001170902.1 | |
| NM_001177433.1 | 8 | 1487 | NP_001170904.1 | |
| NM_021625.4 | 11 | 1866 | NP_067638.3 | |
| NM_147204.2 | 9 | 1628 | NP_671737.1 | |
| XM_005253918.1 | 11 | 2226 | XP_005253975.1 | |
| XM_011538630.2 | 11 | 2969 | XP_011536932.2 | |
| XM_011538631.2 | 10 | 2829 | XP_011536933.2 | |
| XM_011538632.2 | 10 | 2790 | XP_011536934.2 | |
| XM_011538633.2 | 9 | 2649 | XP_011536935.2 | |
| XM_011538634.2 | 11 | 2971 | XP_011536936.2 | |
| XM_011538635.2 | 11 | 2973 | XP_011536937.1 | |
| XM_011538636.2 | XP_011536938.1 | |||
| XM_017019774.1 | 11 | 2599 | XP_016875263.1 | |
| AB032427.1 | 10 | 1872 | BAB69040.1 | |
| AB073669.1 | 9 | 1597 | BAC06573.1 | |
| AB100308.1 | 10 | 1777 | BAC55864.1 | |
| AF258465.1 | 10 | 1777 | AAG16127.1 | |
| AF263523.1 | 10 | 1777 | AAG28029.1 | |
| AF279673.1 | 10 | 1573 | AAK69487.1 | |
| AJ296305.1 | 11 | 1866 | CAC82937.1 | |
| AL834192.1 | 11 | 1837 | ||
| BC117426.1 | 11 | 1860 | ||
| BC143315.1 | 11 | 1860 | ||
| DQ059644.1 | 9 | 1636 | AAZ04918.1 | |
| DQ059645.1 | 11 | 1675 | AAZ04919.1 | |
| DQ059646.1 | 8 | 1456 | AAZ04920.1 |
More Information
Set Membership: |
|

Genomic Map