Assay Details
Target Gene Details
Entrez Gene ID: | 5660 |
Gene Name: | prosaposin |
Gene Aliases: |
GLBA, SAP1 |
Location: |
Chr.10:71816283-71851375 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSAP | NM_001042465.2 | 15 | 2441 | NP_001035930.1 |
| NM_001042466.2 | 15 | 2438 | NP_001035931.1 | |
| NM_002778.3 | 14 | 2432 | NP_002769.1 | |
| AA487963.1 | ||||
| AK057878.1 | ||||
| AK129790.1 | ||||
| BC001503.2 | 14 | 2283 | AAH01503.1 | |
| BC004275.1 | 14 | 2291 | AAH04275.1 | |
| BC007612.1 | 14 | 2296 | AAH07612.1 | |
| BC091483.1 | 9 | 1478 | AAH91483.1 | |
| D00422.1 | 15 | 2295 | BAA00321.1 | |
| J03015.1 | 14 | 2284 | AAB59494.1 | |
| J03077.1 | 14 | 2316 | AAA52560.1 | |
| M60255.1 | 15 | 2287 | AAA36594.1 | |
| M60257.1 | 14 | 2278 | AAA36595.1 | |
| M60258.1 | 15 | 2284 | AAA36596.1 | |
| M81355.1 | 15 | 2295 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831910 | Chr.10:71635558 - 71843223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map