Assay Details
Target Gene Details
Entrez Gene ID: | 390162 |
Gene Name: | olfactory receptor family 5 subfamily M member 9 |
Gene Aliases: |
OR11-190 |
Location: |
Chr.11:56462469-56463401 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR5M9 | NM_001004743.1 | NP_001004743.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1948n54 | Chr.11:56316968 - 56467585 on Build GRCh38 | Loss |
|
| esv3579724 | Chr.11:56310597 - 56470011 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map