Assay Details
Target Gene Details
Entrez Gene ID: | 152926 |
Gene Name: | protein phosphatase, Mg2+/Mn2+ dependent 1K |
Gene Aliases: |
BDP, MSUDMV, PP2Ckappa, PP2Cm, PTMP, UG0882E07 |
Location: |
Chr.4:88257609-88284831 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPM1K | NM_152542.4 | 7 | 1655 | NP_689755.3 |
| XM_006714111.3 | 7 | 1716 | XP_006714174.1 | |
| XM_017007803.1 | 7 | 1303 | XP_016863292.1 | |
| XM_017007804.1 | 8 | 1843 | XP_016863293.1 | |
| XM_017007805.1 | 9 | 1843 | XP_016863294.1 | |
| XM_017007806.1 | 7 | 1176 | XP_016863295.1 | |
| XM_017007807.1 | 6 | 894 | XP_016863296.1 | |
| AK054678.1 | 7 | 1383 | BAB70790.1 | |
| AL834271.1 | 7 | 1374 | CAD38946.1 | |
| AY157615.1 | 7 | 1383 | AAO17296.1 | |
| AY435431.1 | 7 | 1560 | AAR06213.1 | |
| AY994097.1 | 7 | 1391 | AAX77016.1 | |
| BC037552.1 | 7 | 1419 | AAH37552.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759266 | Chr.4:87997399 - 88291967 on Build GRCh38 | Gain |
|
| esv3575758 | Chr.4:88250298 - 88318857 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map