Assay Details
Target Gene Details
Entrez Gene ID: | 54206 |
Gene Name: | ERBB receptor feedback inhibitor 1 |
Gene Aliases: |
GENE-33, MIG-6, MIG6, RALT |
Location: |
Chr.1:8011719-8026333 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERRFI1 | NM_018948.3 | NP_061821.1 | ||
| XM_005263477.2 | XP_005263534.1 | |||
| XM_006710697.2 | XP_006710760.1 | |||
| XM_011541596.2 | XP_011539898.1 | |||
| AJ276373.1 | CAC20426.1 | |||
| AK096149.1 | ||||
| AK315718.1 | ||||
| BC025337.1 | AAH25337.1 | |||
| DB033666.1 |
More Information
Set Membership: |
|

Genomic Map