Assay Details
Target Gene Details
Entrez Gene ID: | 2644 |
Gene Name: | GTP cyclohydrolase I feedback regulator |
Gene Aliases: |
GFRP, HsT16933, P35 |
Location: |
Chr.15:40764087-40767713 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCHFR | NM_005258.2 | 3 | 336 | NP_005249.1 |
| AK223430.1 | 3 | 355 | BAD97150.1 | |
| AK311872.1 | ||||
| BC112262.1 | 3 | 296 | AAI12263.1 | |
| BC112264.1 | 3 | 296 | AAI12265.1 | |
| BC143904.1 | 3 | 263 |
More Information
Set Membership: |
|

Genomic Map