Assay Details
Target Gene Details
Entrez Gene ID: | 32 |
Gene Name: | acetyl-CoA carboxylase beta |
Gene Aliases: |
ACC2, ACCB, HACC275 |
Location: |
Chr.12:109112666-109268226 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACACB | NM_001093.3 | 1 | 621 | NP_001084.3 |
| XM_005253876.4 | 2 | 731 | XP_005253933.1 | |
| XM_011538259.2 | 2 | 1369 | XP_011536561.1 | |
| XM_011538263.2 | 2 | 1369 | XP_011536565.1 | |
| XM_011538264.2 | XP_011536566.1 | |||
| XM_011538265.2 | 2 | 1371 | XP_011536567.1 | |
| AJ575431.2 | 1 | 612 | CAE01470.2 | |
| AJ575592.3 | 1 | 612 | CAE01471.3 | |
| AY382667.1 | 2 | 731 | AAR37018.1 | |
| DQ493870.1 | 1 | 612 | ||
| U89344.1 | 1 | 612 | AAB58382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528323 | Chr.12:109115044 - 109225776 on Build GRCh38 | Gain |
|
| dgv1553n100 | Chr.12:109064624 - 109159352 on Build GRCh38 | Gain |
|
| nsv526593 | Chr.12:109131263 - 109140147 on Build GRCh38 | Loss |
|
| dgv1554n100 | Chr.12:109104860 - 109232663 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map