Assay Details
Target Gene Details
Entrez Gene ID: | 1263 |
Gene Name: | polo like kinase 3 |
Gene Aliases: |
CNK, FNK, PLK-3, PRK |
Location: |
Chr.1:44799952-44805995 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLK3 | NM_004073.3 | 14 | 1922 | NP_004064.2 |
| AA421212.1 | ||||
| AB209875.1 | 15 | 1609 | BAD93112.1 | |
| AJ293866.1 | 14 | 1922 | CAC10659.1 | |
| BC013899.2 | 14 | 1783 | AAH13899.1 | |
| U56998.1 | 14 | 1602 | AAC50637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950705 | Chr.1:44801029 - 44813128 on Build GRCh38 | Deletion |
|
| nsv546145 | Chr.1:44752870 - 44827846 on Build GRCh38 | Loss |
|
| nsv546147 | Chr.1:44799202 - 44805425 on Build GRCh38 | Loss |
|
| nsv546146 | Chr.1:44792469 - 44853080 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map