Assay Details
Target Gene Details
Entrez Gene ID: | 51102 |
Gene Name: | mitochondrial trans-2-enoyl-CoA reductase |
Gene Aliases: |
CGI-63, FASN2B, NRBF1 |
Location: |
Chr.1:29192657-29230958 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MECR | NM_001024732.2 | 10 | 1625 | NP_001019903.2 |
| NM_016011.3 | 10 | 1498 | NP_057095.3 | |
| XM_005245885.2 | 11 | 1548 | XP_005245942.1 | |
| AK095099.1 | 10 | 1591 | ||
| AK225687.1 | 10 | 1464 |
More Information
Set Membership: |
|

Genomic Map