Assay Details
Target Gene Details
Entrez Gene ID: | 79018 |
Gene Name: | GID complex subunit 4 homolog |
Gene Aliases: |
C17orf39, VID24 |
Location: |
Chr.17:18039297-18068405 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GID4 | NM_024052.4 | 6 | 2944 | NP_076957.3 |
| XM_005256800.1 | 5 | 2700 | XP_005256857.1 | |
| AK124377.1 | ||||
| AK127580.1 | 6 | 2631 |
More Information
Additional Information:
For this assay, SNP(s) [rs76619863] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map