Assay Details
Target Gene Details
Entrez Gene ID: | 26470 |
Gene Name: | seizure related 6 homolog like 2 |
Gene Aliases: |
BSRPA, PSK-1 |
Location: |
Chr.16:29871159-29899573 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEZ6L2 | NM_001114099.2 | 15 | 2588 | NP_001107571.1 |
| NM_001114100.2 | 14 | 2495 | NP_001107572.1 | |
| NM_001243332.1 | 16 | 2837 | NP_001230261.1 | |
| NM_001243333.1 | 15 | 2705 | NP_001230262.1 | |
| NM_012410.3 | 16 | 2627 | NP_036542.1 | |
| NM_201575.3 | 15 | 2798 | NP_963869.2 | |
| XM_005255252.2 | 15 | 2645 | XP_005255309.1 | |
| XM_017023135.1 | 14 | 2594 | XP_016878624.1 | |
| AB105376.1 | 16 | 2466 | ||
| AF131749.1 | 8 | 1358 | ||
| AJ245820.1 | 16 | 2466 | CAB57950.1 | |
| AJ245821.1 | 16 | 2466 | CAB57951.1 | |
| AJ245822.1 | 15 | 2427 | CAB57952.1 | |
| AK027407.1 | 14 | 2107 | ||
| AK027676.1 | 13 | 2077 | ||
| AK074998.1 | 12 | 1795 | BAC11345.1 | |
| AK293675.1 | 15 | 2550 | ||
| AK299103.1 | 16 | 2834 | ||
| AY358404.1 | 15 | 2787 | AAQ88770.1 | |
| BC000567.2 | 14 | 2295 | AAH00567.1 | |
| KJ534938.1 | 13 | 2061 | ||
| KJ534939.1 | 13 | 2217 | ||
| KJ534940.1 | 15 | 2391 | ||
| KJ534941.1 | 14 | 2250 | ||
| KJ534988.1 | 14 | 2262 | ||
| KJ534989.1 | 5 | 592 | ||
| KJ535068.1 | 14 | 2110 | ||
| KJ535092.1 | 16 | 2591 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571818 | Chr.16:29861040 - 29874126 on Build GRCh38 | Loss |
|

Genomic Map