Assay Details
Target Gene Details
Entrez Gene ID: | 192666 |
Gene Name: | keratin 24 |
Gene Aliases: |
K24, KA24 |
Location: |
Chr.17:40694246-40703750 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT24 | NM_019016.2 | NP_061889.2 | ||
| XM_006721739.3 | XP_006721802.1 | |||
| XM_011524460.2 | XP_011522762.1 | |||
| XM_017024299.1 | XP_016879788.1 | |||
| AK000268.1 | BAA91044.1 | |||
| BC111968.1 | AAI11969.1 |
More Information
Set Membership: |
|

Genomic Map