Assay Details
Target Gene Details
Entrez Gene ID: | 79748 |
Gene Name: | lectin, mannose binding 1 like |
Gene Aliases: |
ERGIC-53L, ERGL |
Location: |
Chr.15:74812853-74825758 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMAN1L | NM_021819.2 | NP_068591.2 | ||
| AF303398.1 | AAG30902.1 | |||
| AK291819.1 | ||||
| AK294807.1 | ||||
| AK300517.1 | ||||
| AK309976.1 | ||||
| AY358724.1 | AAQ89086.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833055 | Chr.15:74696003 - 74873947 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112360380] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map