Assay Details
Target Gene Details
Entrez Gene ID: | 57194 |
Gene Name: | ATPase phospholipid transporting 10A (putative) |
Gene Aliases: |
ATP10C, ATPVA, ATPVC |
Location: |
Chr.15:25672241-25865144 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP10A | NM_024490.3 | 21 | 5049 | NP_077816.1 |
| XM_005268261.4 | 22 | 5179 | XP_005268318.1 | |
| XM_011521826.2 | 22 | 5350 | XP_011520128.1 | |
| XM_011521828.2 | XP_011520130.1 | |||
| XM_011521829.2 | 18 | 4182 | XP_011520131.1 | |
| XM_011521830.2 | 16 | 3852 | XP_011520132.2 | |
| XM_017022436.1 | 21 | 4685 | XP_016877925.1 | |
| XM_017022437.1 | 21 | 5049 | XP_016877926.1 | |
| AB011138.1 | 16 | 3937 | BAA25492.1 | |
| AB051358.1 | 22 | 5142 | BAB47392.1 | |
| BC052251.1 | 21 | 5055 | AAH52251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569331 | Chr.15:25376733 - 26051249 on Build GRCh38 | Gain |
|
| nsv483024 | Chr.15:25540816 - 25708423 on Build GRCh38 | Loss |
|
| nsv568586 | Chr.15:25678071 - 25691886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map