Assay Details
Target Gene Details
Entrez Gene ID: | 525 |
Gene Name: | ATPase H+ transporting V1 subunit B1 |
Gene Aliases: |
ATP6B1, RTA1B, VATB, VMA2, VPP3 |
Location: |
Chr.2:70935868-70965431 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1B1 | NM_001692.3 | NP_001683.2 | ||
| XM_011532907.2 | XP_011531209.1 | |||
| AK127853.1 | ||||
| AK223151.1 | BAD96871.1 | |||
| AK291121.1 | ||||
| AK301542.1 | ||||
| AK313194.1 | ||||
| AU126647.2 | ||||
| BC063411.1 | AAH63411.1 | |||
| DA845716.1 | ||||
| M25809.1 | AAA36498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013232 | Chr.2:70849342 - 71049568 on Build GRCh38 | Gain |
|
| nsv1014516 | Chr.2:70881638 - 70962921 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map