Assay Details
Target Gene Details
Entrez Gene ID: | 64766 |
Gene Name: | S100P binding protein |
Gene Aliases: |
S100PBPR |
Location: |
Chr.1:32816767-32858879 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100PBP | XM_011541962.2 | XP_011540264.1 |
Target Gene Details
Entrez Gene ID: | 8565 |
Gene Name: | tyrosyl-tRNA synthetase |
Gene Aliases: |
CMTDIC, TYRRS, YRS, YTS |
Location: |
Chr.1:32775237-32818032 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YARS | NM_003680.3 | 1 | 699 | NP_003671.1 |
| XM_011542347.1 | XP_011540649.1 | |||
| XM_011542348.1 | 1 | 42 | XP_011540650.1 | |
| XM_017002651.1 | XP_016858140.1 | |||
| AK026535.1 | ||||
| AK125213.1 | 1 | 820 | ||
| AK223608.1 | BAD97328.1 | |||
| BC016689.1 | AAH16689.1 |
More Information
Set Membership: |
|

Genomic Map