Assay Details
Target Gene Details
Entrez Gene ID: | 89801 |
Gene Name: | protein phosphatase 1 regulatory subunit 3F |
Gene Aliases: |
HB2E, LL0XNC01-7P3.1, R3F |
Location: |
Chr.X:49269822-49301465 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R3F | NM_001184745.1 | 4 | 1171 | NP_001171674.1 |
| NM_033215.4 | 4 | 2114 | NP_149992.3 | |
| XM_005272687.3 | 4 | 2132 | XP_005272744.1 | |
| XM_017029932.1 | 4 | 6542 | XP_016885421.1 | |
| XM_017029933.1 | 4 | 6539 | XP_016885422.1 | |
| XM_017029934.1 | XP_016885423.1 | |||
| XM_017029935.1 | XP_016885424.1 | |||
| AK056297.1 | 3 | 1531 | ||
| AK056909.1 | 4 | 1939 | ||
| AK127066.1 | 1 | 2771 | BAC86811.1 | |
| AK303738.1 | 5 | 1437 | ||
| BC110405.1 | 1 | 699 | ||
| BC131588.1 | 4 | 1144 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain |
|
| nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map