Assay Details
Target Gene Details
Entrez Gene ID: | 80179 |
Gene Name: | myosin XIX |
Gene Aliases: |
MYOHD1 |
Location: |
Chr.17:36495636-36535457 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO19 | NM_001163735.1 | NP_001157207.1 | ||
| NM_025109.5 | NP_079385.2 | |||
| XM_011525286.1 | XP_011523588.1 | |||
| XM_011525287.1 | XP_011523589.1 | |||
| XM_011525290.2 | XP_011523592.1 | |||
| XM_017025157.1 | XP_016880646.1 | |||
| XM_017025158.1 | XP_016880647.1 | |||
| XM_017025159.1 | XP_016880648.1 | |||
| XM_017025160.1 | XP_016880649.1 | |||
| XM_017025161.1 | XP_016880650.1 | |||
| AB209035.1 | BAD92272.1 | |||
| AK293903.1 | ||||
| AK304073.1 | ||||
| AL133017.1 | ||||
| BC008900.2 | AAH08900.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640472 | Chr.17:36483375 - 36567136 on Build GRCh38 | Gain |
|
| esv2763162 | Chr.17:36460442 - 36567250 on Build GRCh38 | Gain |
|
| esv3640471 | Chr.17:36461662 - 36600489 on Build GRCh38 | Gain |
|
| nsv469853 | Chr.17:36387554 - 36574537 on Build GRCh38 | Gain+Loss |
|
| nsv471697 | Chr.17:36357257 - 36574537 on Build GRCh38 | Gain |
|
| nsv516100 | Chr.17:36459737 - 36549567 on Build GRCh38 | Gain+Loss |
|
| esv3640473 | Chr.17:36507699 - 36538931 on Build GRCh38 | Gain |
|
| dgv573e214 | Chr.17:36461232 - 36515958 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map