Assay Details
Target Gene Details
Entrez Gene ID: | 10411 |
Gene Name: | Rap guanine nucleotide exchange factor 3 |
Gene Aliases: |
CAMP-GEFI, EPAC, EPAC1, HSU79275, bcm910 |
Location: |
Chr.12:47734670-47759106 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAPGEF3 | NM_001098531.2 | NP_001092001.1 | ||
| NM_001098532.2 | NP_001092002.1 | |||
| NM_006105.5 | NP_006096.2 | |||
| XM_005268571.3 | XP_005268628.1 | |||
| XM_011537752.1 | XP_011536054.1 | |||
| XM_011537755.1 | XP_011536057.1 | |||
| XM_011537758.2 | XP_011536060.1 | |||
| AF103905.1 | AAC83381.1 | |||
| AK055502.1 | ||||
| AK074444.1 | ||||
| AK092448.1 | ||||
| AK125545.1 | ||||
| AK290230.1 | ||||
| AK299029.1 | ||||
| BC017728.1 | ||||
| BC092404.1 | ||||
| BQ007070.1 | ||||
| U78168.1 | AAD12740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv696 | Chr.12:47720180 - 47754753 on Build GRCh38 | Insertion |
|
| nsv470289 | Chr.12:47716768 - 47786725 on Build GRCh38 | Gain |
|
| dgv94n21 | Chr.12:47725779 - 47821397 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map