Assay Details
Target Gene Details
Entrez Gene ID: | 31 |
Gene Name: | acetyl-CoA carboxylase alpha |
Gene Aliases: |
ACAC, ACACAD, ACC, ACC1, ACCA |
Location: |
Chr.17:37084992-37406822 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACACA | NM_198834.2 | NP_942131.1 | ||
| NM_198839.2 | NP_942136.1 | |||
| XM_006721853.1 | XP_006721916.1 | |||
| XM_011524701.1 | XP_011523003.1 | |||
| XM_017024553.1 | XP_016880042.1 | |||
| XM_017024554.1 | XP_016880043.1 | |||
| AK308905.1 | ||||
| AY315623.1 | AAP94118.1 | |||
| AY315625.1 | AAP94120.1 | |||
| AY315626.1 | AAP94121.1 | |||
| BC137287.1 | ||||
| BF154865.1 | ||||
| CF454424.1 |
Target Gene Details
Entrez Gene ID: | 284099 |
Gene Name: | chromosome 17 open reading frame 78 |
Gene Aliases: |
- |
Location: |
Chr.17:37375710-37392708 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C17orf78 | NM_001321399.1 | NP_001308328.1 | ||
| NM_173625.4 | NP_775896.3 | |||
| XM_011524647.2 | XP_011522949.1 | |||
| XM_011524648.2 | XP_011522950.1 | |||
| XM_011524649.2 | XP_011522951.1 | |||
| AK096966.1 | BAC04914.1 | |||
| BC034672.1 | AAH34672.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32692 | Chr.17:37378162 - 37398642 on Build GRCh38 | Loss |
|
| esv3640487 | Chr.17:37198366 - 37397715 on Build GRCh38 | Gain |
|
| dgv3157n100 | Chr.17:37219334 - 37462210 on Build GRCh38 | Gain |
|
| nsv574928 | Chr.17:37387113 - 37398642 on Build GRCh38 | Loss |
|
| esv2758687 | Chr.17:37211817 - 37414232 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map