Assay Details
Target Gene Details
Entrez Gene ID: | 56952 |
Gene Name: | phosphoribosyl transferase domain containing 1 |
Gene Aliases: |
HHGP |
Location: |
Chr.10:24848607-24952644 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRTFDC1 | NM_001282786.1 | 8 | 687 | NP_001269715.1 |
| NM_020200.6 | 9 | 764 | NP_064585.1 | |
| AB209792.1 | 9 | 714 | BAD93029.1 | |
| AF226056.1 | 9 | 712 | AAF86956.1 | |
| AK021950.1 | 9 | 724 | BAB13944.1 | |
| AK222674.1 | 9 | 723 | BAD96394.1 | |
| AK294130.1 | 8 | 646 | ||
| BC008662.2 | 9 | 726 | AAH08662.1 |
More Information
Set Membership: |
|

Genomic Map